Learn More
Medchemexpress LLC Doramectin | 117704-25-3 | 97.4% | 899.11 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY17035R50MG
Encompass_Preferred
Doramectin (Standard) is the analytical standard of Doramectin, intended for research and analytical applications. It is a derivative of Ivermectin and is described as a potent antiparasitic antibiotic. Doramectin is an active compound against S. mansoni in an NMRI mouse infection model. This analytical standard is commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.
  • Intended for research and analytical applications
  • Potent antiparasitic antibiotic
  • Active compound against S. mansoni in an NMRI mouse infection model
  • Used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS
Catalog No. 50-003-84064