Learn More
Medchemexpress LLC Doramectin | 117704-25-3 | MFCD00894747 | >95.0% | 10 MG
SDP
Supplier:  Medchemexpress LLC HY17035R10MG
Encompass_Preferred
Doramectin (Standard) is the analytical reference standard of doramectin, a macrocyclic lactone antiparasitic derivative of ivermectin. It is provided for research and analytical applications and is characterized by CAS 117704-25-3, molecular formula C50H74O14, and molecular weight 899.11. The material is supplied in solid form and as a solution for use in assay, calibration, and method development.
  • Analytical standard suitable for assay and method development.
  • Characterized by CAS 117704-25-3 and molecular weight 899.11.
  • Available as solid and DMSO solution formats for flexible use.
  • Intended for research and analytical applications.
  • Certificate of analysis available; confirm supplier-specific purity before use.
Catalog No. 50-002-95432