Learn More
Apexbio Technology LLC Doramectin,5g CAS# 117704-25-3
SDP
Supplier:  Apexbio Technology LLC A33715000
Encompass_Preferred
Doramectin (Dectomax) is a veterinary drug approved by the Food and Drug Administration (FDA) for the treatment of parasites such as gastrointestinal roundworms, lungworms, eyeworms, grubs, sucking lice and mange mites in cattle. Mutational biosynthetic antiparasitic antibiotic structurally related to the avermectins. Other sizes are also available. Please inqury us for quote.
Catalog No. 50-224-0705