Learn More
Medchemexpress LLC Doramectin (Standard) | 117704-25-3 | 97.4% | 899.11 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY17035R100MG
Encompass_Preferred
Doramectin (Standard) is the analytical standard of Doramectin, intended for research and analytical applications. It is a derivative of Ivermectin and a potent antiparasitic antibiotic active against *S. mansoni* in an NMRI mouse infection model. This product is for research use only.
  • Used as a reference standard for assays
  • Commonly utilized in qualitative, quantitative, and methodological research experiments
  • Applicable in HPLC, GC, and MS
Catalog No. 50-003-84497