Learn More
Medchemexpress LLC Doramectin (Standard) | 117704-25-3 | 97.4% | 899.11 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY17035R
Encompass_Preferred
Doramectin (Standard) is the analytical standard of Doramectin, intended for research and analytical applications. It is a derivative of Ivermectin and acts as a potent antiparasitic antibiotic. This compound is active against *S. mansoni* in an NMRI mouse infection model and is used as a reference for various assays.
  • Analytical standard for research and analytical applications.
  • Derivative of Ivermectin.
  • Potent antiparasitic antibiotic.
  • Active against *S. mansoni* in NMRI mouse infection model.
  • Used in qualitative, quantitative, and methodological research experiments.
  • Suitable for HPLC, GC, and MS applications.
Catalog No. 50-003-85130