Learn More
Apexbio Technology LLC Doripenem 148016-81-3 100mg
SDP
Supplier:  Apexbio Technology LLC B1076100
Encompass_Preferred
Doripenem (148016-81-3) is a small-molecule inhibitor targeting penicillin-binding proteins (PBPs) It is designed to inhibit bacterial cell wall peptidoglycan synthesis thereby impairing cell wall integrity Doripenem exerts its biological activity primarily through inhibition of bacterial cell wall synthesis by binding to essential PBPs In in vitro studies Doripenem demonstrates inhibitory activity with IC50 values typically in the low micromolar range against representative bacterial isolates Based on these pharmacological properties Doripenem holds research potential in investigating microbial resistance mechanisms evaluating beta-lactamase enzyme inhibition and studying pharmacokinetic/pharmacodynamic profiles relevant to antimicrobial development
Catalog No. 50-000-89572