Learn More
Alkali Scientific Doripenem, 250 Mg
SDP
Supplier:  Alkali Scientific DOR002CP
Encompass_Preferred
Doripenem is a potent carbapenem antibiotic used in laboratory research to investigate bacterial resistance and the effectiveness of antimicrobials. The 250mg dose is used for testing in microbiological studies, including bacterial culture and antibiotic susceptibility assays. Doripenem works by inhibiting bacterial cell wall synthesis, which makes it highly effective against a broad spectrum of bacteria. This compound is essential for studying the mechanisms of resistance, particularly in pathogens resistant to other classes of antibiotics. The product is widely used in clinical microbiology and pharmaceutical research, helping to develop strategies for combating multi-drug resistant bacteria.
Catalog No. 50-006-60168