Learn More
Medchemexpress LLC Dorsomorphin | 866405-64-3 | 99.10% | 100 MG
SDP
Supplier:  Medchemexpress LLC 100MGHY13418A
Encompass_Preferred
Dorsomorphin (Compound C) is a selective and ATP-competitive AMPK inhibitor. It selectively inhibits BMP type I receptors ALK2, ALK3, and ALK6. Additionally, it can reverse autophagy activation and the anti-inflammatory effect of Urolithin A.
  • Selective and ATP-competitive AMPK inhibitor (Ki of 109 nM in the absence of AMP)
  • Selectively inhibits BMP type I receptors ALK2, ALK3, and ALK6
  • Can reverse autophagy activation
  • Can reverse the anti-inflammatory effect of Urolithin A
Catalog No. 50-003-64711