Learn More
Apexbio Technology LLC Dorsomorphin (Compound C) 866405-64-3 50mg
SDP
Supplier:  Apexbio Technology LLC B325250
Encompass_Preferred
Dorsomorphin (Compound C CAS 866405-64-3) is a cell-permeable reversible ATP-competitive inhibitor of AMP-activated protein kinase (AMPK) exhibiting a Ki value of 109 nM It displays high specificity towards AMPK relative to related kinases such as protein kinase A protein kinase C and Janus kinase 3 Additionally dorsomorphin acts as an inhibitor of BMP signaling by reducing phosphorylation of Smad1/5/8 leading to inhibited osteogenic differentiation elevated iron levels via decreased hepcidin transcription and enhanced human ESC self-renewal and neural induction
Catalog No. 50-000-63907