Learn More
Selleck Chemical LLC Dorsomorphin (Compound C) 5mg 866405-64-3 BML-275,Compound C
SDP
Supplier:  Selleck Chemical LLC S78405MG
Encompass_Preferred
Dorsomorphin (Compound C, BML-275) is a potent, reversible, selective AMPK inhibitor with Ki of 109 nM in cell-free assays, exhibiting no significant inhibition of several structurally related kinases including ZAPK, SYK, PKC?, PKA, and JAK3. Dorsomorphin selectively inhibits the BMP type I receptors ALK2, ALK3 and ALK6. Dorsomorphin is used in promoting specific cell differentiation and inducing cancer cell line autophagy. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-5325