Learn More
Medchemexpress LLC Dorzagliatin | 1191995-00-2 | 99.89% | 462.93 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY109030100MG
Encompass_Preferred
Dorzagliatin (HMS5552) is a dual-acting glucokinase (GK) activator that improves glycaemic control and pancreatic β-cell function in type 2 diabetes. It is for research use only and not sold to patients.
  • Dual-acting glucokinase (GK) activator
  • Improves glycaemic control and pancreatic β-cell function in type 2 diabetes
Catalog No. 50-003-46574