Learn More
Medchemexpress LLC Dorzagliatin | 1191995-00-2 | MFCD22376579 | 98.5% | 462.93 g·mol-1 | C22H27ClN4O5 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1090305MG
Encompass_Preferred
Dorzagliatin is a dual-acting glucokinase (GK) activator supplied as a research standard for biochemical and pharmacological studies. It modulates glucose sensing and pancreatic β-cell function and is commonly used in preclinical diabetes research and mechanism-of-action studies. Not for human or clinical use.
Catalog No. 50-002-64235