Learn More
BURKLE INC Dosing pump Dosi-Pump, 3.38 oz. (100 ml)stroke, fix. feed.tube
Supplier:  BURKLE INC 56074100
Encompass_Preferred
Dosing pumps are mounted by you in your existing container closure (see the locating bore table). To do this, a suitable hole is drilled in the cap. Then the pump is inserted through the hole and screwed in place from below.
A matching container connection is not included in the scope of delivery.
Catalog No. 50-000-25902