Learn More
Labelmaster Inc Special Permit Packaging DOT-SP-9168, 950 CC Bottle for Poison Inhalation Hazard
SDP
Supplier:  Labelmaster Inc HMS950PIH
Encompass_Preferred
This fully assembled Special Permit Packaging, DOT-SP 9168, 950 cc bottle kit can be used to ship Packing Groups I, II or III materials by all modes of transport (see Special Permit for details). The exemption packages can be used to ship solids or liquid materials with a specific gravity of 2.0 or less. Special Permit Packaging DOT-SP 9168 meets Procedure 1A requirements and is accepted by UPS and FedEx as an exemption package. The DOT-SP 9168 kits carry an exemption that allows you to ship Hazard Classes 3, 4, 5, 6, 8 and 9 domestically without using a hazard class label.
Catalog No. NC1921205