Learn More
Medchemexpress LLC HY-135127 5mg Medchemexpress, Dot1L-IN-4 CAS:2565705-02-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1351275MG
Encompass_Preferred
Medchemexpress, HY-135127 5mg Dot1L-IN-4 CAS:2565705-02-2 Dot1L-IN-4 is a potent disruptor of telomeric silencing 1-like protein (DOT1L) inhibitor with an IC50 SPA DOT1L of 0.11 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1379