Learn More
Carbosynth LLC. DOTA-Glu-[Cyclo(Arg-Gly-Asp-D-Phe-Lys)]2, 5mg, MW: 1704.88, Molecular formula: C75H113N23O23
SDP
Supplier:  Carbosynth LLC. PCI3903PI5MG
Encompass_Preferred
Synonyms: DOTA-Glu-[cyclo(Arg-Gly-Asp-D-Phe-Lys)]2, DOTA-E-[c(RGDfK)2], Application: αvß3 Integrin Binding RGD Peptide RGD Tumor Targeting Peptide for Radiolabelling (Requires further derivatization before use), Storage Temperature: -20°C, References: M.L. Janssen, W.J. Oyen, I. Dijkgraaf, L.F. Massuger, C. Frielink, D.S. Edwards, M. Rajopadhye, H. Boonstra, F.H. Corstens, and O.C. Boerman, Cancer Res., 62, 6146 (2002), X. Chen, S. Liu, Y. Hou, M. Tohme, R. Park, J.R. Bading, and P.S. Conti, Mol. Imaging Biol., 6, 350 (2004).
Catalog No. 50-168-6321