Learn More
Medchemexpress LLC DOTA-NOC | 619300-53-7 | 99.2% | 1455.70 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYP21121MG
Encompass_Preferred
DOTA-NOC (DOTA-Nal3-octreotide) is a high-affinity ligand for somatostatin receptor subtypes 2, 3, and 5. This compound is valuable for labeling with various radiometals and for advancing radiopeptide imaging techniques. It also serves as a key component in the synthesis and research of Radionuclide-Drug Conjugates (RDCs).
  • High-affinity ligand for somatostatin receptor subtypes 2, 3, and 5
  • Can be used for labeling with various radiometals
  • Suitable for the development of radiopeptide imaging
  • Utilized for the synthesis and research of radionuclide-drug conjugates (RDCs)
Catalog No. 50-004-24353