Learn More
Cayman Chemical DOTA-NOCacetate 5mg
SDP
Supplier:  Cayman Chemical 419755 MG
Encompass_Preferred
A synthetic peptide; has been conjugated to radioligands for use in PET imaging of neuroendocrine tumors and for tissue distribution studies in vivo
Catalog No. 50-294-3336