Learn More
CPC Scientific DOTA-DPhe-Cys-Phe-DTrp-Lys-Thr-Cys-Thr-ol (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific SOMA015B
Encompass
A group of compounds comprising DOTA, a metal chelator that carries a radionuclide and octreotide (an octapeptide that is a potent somatostatin (SST)) analog (where DOTA is tetra-aza-cyclododecane-N N'N"N-"'-tetra-acetate). ONE-LETTER SEQUENCE: DOTA-fCFwKTCT-ol (C2&C7 bridge)MOLECULAR FORMULA: C65H92N14O17S2MOLECULAR WEIGHT:1405.7STORAGE CONDITIONS: -20 5CRESEARCH AREA: GastrointestinalREFERENCES: L.Kolby et al., Br. J. Cancer, 93, 1144 (2005); R.Valkema et al., J. Nucl. Med., 46, 83S (2005); M.de Jong et al., J. Nucl. Med., 46, 13S (2005); W.A.Breeman et al., Eur. J. Nucl. Med. Mol. Imaging, 30, 917 (2003); A.Capello et al., Cancer Biother. Radiopharm., 18, 761 (2003); Chinol, M. et al. Semin. Nucl. Med. 32,141(2002); De Jong, M. et al. semin Nucl. Med. 32,133(2002)
Catalog No. 50-211-0440