Learn More
Medchemexpress LLC Dota-PEG5-amine | 00-00-0 | 98.0% | C28H54N6O12 | 10MG
SDP
Supplier:  Medchemexpress LLC HY14000810MG
Encompass_Preferred
DOTA-PEG5-amine is a polyethylene glycol (PEG)-based linker incorporating a DOTA chelator and a terminal primary amine. It is intended for use in the synthesis of PROTACs and other conjugates where a chelation site and a reactive amine are required; supplied as a white to off-white solid, characterized by molecular formula C28H54N6O12 and molecular weight 666.76 g·mol⁻¹.
  • PEG-based linker with a terminal amine.
  • Contains a DOTA chelator suitable for metal ion binding and radiolabeling.
  • High purity (98.0%) appropriate for synthetic applications.
  • Available in multiple small pack sizes for research use.
  • Stable as a powder with defined storage conditions for long-term and short-term storage.
Catalog No. 50-002-58488