Learn More
Medchemexpress LLC DOTA-PEG5-amine | 98.0% | 666.76 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY140008100MG
Encompass_Preferred
DOTA-PEG5-amine is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. These compounds leverage the intracellular ubiquitin-proteasome system to selectively degrade target proteins and are intended for research use only.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Composed of two distinct ligands joined by a linker
  • One ligand targets an E3 ubiquitin ligase
  • Other ligand targets a specific protein
  • Leverages the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
Catalog No. 50-003-50787