Learn More
Medchemexpress LLC Dota-PEG5-azide | 00-00-0 | 98.0% | C28H52N8O12 | 10MG
SDP
Supplier:  Medchemexpress LLC HY14075210MG
Encompass_Preferred
DOTA-PEG5-azide is a PEG-based linker that combines a DOTA chelator with an azide functional group for click-chemistry conjugation. Supplied as a white to off-white solid, it is intended for use in PROTAC synthesis, bioconjugation, and radiolabeling workflows where a chelator and a click-reactive handle are required.
  • Contains DOTA chelator suitable for metal chelation and radiolabeling.
  • Azide functional group enables copper-catalyzed and strain-promoted click reactions.
  • PEG5 spacer improves solubility and reduces steric hindrance in conjugates.
  • High purity (98.0%) for research-grade synthesis.
  • Available in small-scale quantities compatible with iterative synthesis workflows.
Catalog No. 50-002-58941