Learn More
Medchemexpress LLC HY-131326 250mg Medchemexpress, DOTA-tris(tBu)ester NHS ester CAS:819869-77-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY131326250MG
Encompass_Preferred
Medchemexpress, HY-131326 250mg DOTA-tris(tBu)ester NHS ester CAS:819869-77-7 DOTA-tris(tBu)ester NHS ester is a derivative of DOTA for the labeling of peptides and antibodies. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-0256