Learn More
Selleck Chemical LLC DOTAP chloride
SDP
Supplier:  Selleck Chemical LLC S690825MG
Encompass_Preferred
DOTAP chloride (1 2-Dioleoyl-3-trimethylammonium-propane chloride) is an effective cationic lipid widely used as a liposomal transfection reagent DOTAP is also a strong activator of the innate immune system in animal cells and a good candidate as an elicitor for the study of plant immunity
Catalog No. 50-000-02296