Learn More
Medchemexpress LLC Dotap (chloride) | 132172-61-3 | 99.9% | 698.54 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY112754A10MM/1ML
Encompass_Preferred
DOTAP chloride is a useful and effective cationic lipid primarily used for transient and stable transfection of DNA (plasmids, bacmids) and modified nucleic acids (antisense oligonucleotides). It facilitates transfection without requiring a helper lipid, with its efficiency being more dependent on lipid composition and cell type rather than size or zeta potential. This product is intended for research use only.
  • Useful for transient and stable transfection of DNA and modified nucleic acids.
  • Does not require a helper lipid for transfection.
  • Transfection efficiency influenced by lipid composition and cell type.
  • For research use only.
  • Solid below 35°C, liquid above 38°C.
  • White to off-white appearance.
Catalog No. 50-004-36943