Learn More
Medchemexpress LLC HY-112754A 250mg , DOTAP (chloride) CAS:132172-61-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY112754A250MG
Encompass_Preferred
HY-112754A 250mg DOTAP (chloride) CAS: 132172-61-3 DOTAP chloride is a useful and effective cationic lipid for transient and stable transfection DNA (plasmids, bacmids) and modified nucleic acids (antisense oligonucleotides) with out the use of helper lipid. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9316