Learn More
Electron Microscopy Sciences Bench Top Double Bin Pipette Work Station
SDP
Supplier:  Electron Microscopy Sciences 6438518
Encompass_Preferred
Pipette work station withclear, hinged acrylic lid, clear acrylic bins for easy viewing, and self-closing lid. Holds 7 pipettors. Clear acrylic lids and fronts permit see-through storage. It may also be used to store small vials, caps, histo-cassettes, aluminum stubs. White. Measures: 18 x 9 x 6.75".
Catalog No. 50-305-80