Learn More
Terumo BCT Inc Double Coupler Adapter
Supplier:  Terumo BCT Inc 90912
Encompass
The double coupler adapter has two spikes that join into one spike port. It gives the operator the flexibility to connect more than one bag of starting cell product to the single spike connector on that line.
Catalog No. NC2005075