Learn More
Health Care Logistics Inc Double-Ended Transfer Needles, 17-Gauge
Supplier: Health Care Logistics Inc 17114
Double-ended 1-1/4" x 3/4" needle. Use for fast transfer of medication from rubber-stoppered vial to vacuumized IV container. 17-GaugeNeedles are individually sterile packaged and encased in a protective sheath. Non-filtered 25 per package