Learn More
Grainger Double Sided Foam Tape 1 1/2 L PK256
Supplier:  Grainger 16U622
Encompass_Preferred
The TAPECASE Double Sided Foam Tape is a white, double-sided adhesive tape made of polyethylene foam. It has an acrylic adhesive and is 0.045 inches thick with a width of 1 inch. The tape can bond a variety of surfaces including wood, plastic, glass, and metal. It has high tack and is designed for permanent bonding for craft, home, office, automotive, and industrial applications. The tape resists UV rays, moisture, and extreme temperatures. It can support up to 5 pounds per square inch when properly applied between clean, dry surfaces. The tape comes in a 33 foot roll and is made in the USA.
Catalog No. 50-276-6956