Learn More
Grainger Double Sided Foam Tape 1/2 L PK256
Supplier:  Grainger 16U612
Encompass_Preferred
The 3M Double Sided Foam Tape 793Z73 is a high performance acrylic adhesive tape with a polyethylene foam carrier. It has a thickness of 1/16 inch and comes in a 36 yard roll. The tape has excellent durability, adhesion strength, and conformability. It bonds well to a variety of surfaces including wood, glass, plastic, and metal. The tape can withstand temperatures from -20F to 200F when applied properly. It has quick stick at room temperature and high initial tack. The tape resists UV light and solvents. It can be used for mounting, bonding, and splicing applications.
Catalog No. 50-276-6053