Learn More
Grainger Double Sided Foam Tape 1 L 1 W PK256
Supplier:  Grainger 16U621
Encompass_Preferred
The 3M Double Sided Foam Tape White is a high-performance acrylic adhesive tape with a polyethylene foam carrier. It has a thickness of 1/16 inch and comes in rolls that are 36 yards long and 1 inch wide. The tape has excellent conformability and bonding strength on a variety of surfaces including wood, glass, plastic, and metal. It can withstand temperatures from -20F to 200F when applied properly. The acrylic adhesive is designed for permanent bonding for both indoor and outdoor applications.
Catalog No. 50-276-6645