Learn More
Grainger Double Sided Foam Tape 2 L 1 W PK162
Supplier:  Grainger 16U611
Encompass_Preferred
The 3M Double Sided Foam Tape is a tan colored double sided adhesive tape made by 3M. It has an acrylic adhesive and comes in multiple widths and lengths. The tape can bond many surfaces including metal, glass, plastic, and wood. It provides high strength bonding and can withstand temperatures up to 200°F. The tape has a thickness of 1/16 inch and can conform to uneven surfaces. It is designed for permanent indoor and outdoor applications.
Catalog No. 50-276-6813