Learn More
Grainger Double Sided Foam Tape 2 L 1 W PK162
Supplier:  Grainger 16U615
Encompass_Preferred
The 3M Double Sided Foam Tape is a tan colored double sided adhesive tape made by 3M. It has an acrylic adhesive and comes in multiple widths and lengths. The tape can bond many materials including metal, glass, plastic, and wood. It provides high strength bonding while allowing for movement due to its conformable foam core. The tape can withstand temperatures from -20°F to 200°F when applied properly. It has quick stick at room temperature and high immediate grab.
Catalog No. 50-276-7527