Learn More
Grainger Double Sided Foam Tape 3/4 L PK256
Supplier:  Grainger 16U624
Encompass_Preferred
The 3M Double Sided Foam Tape 793Z73 is a high performance acrylic adhesive tape with foam carrier. It has a thickness of 0.045 inches and is available on a roll that is 36 yards long and 1 inch wide. This tape features high adhesion levels and can bond a wide range of materials including metals, glass, plastics, and painted surfaces. It can withstand temperatures up to 200°F and has good solvent resistance. The acrylic adhesive resists UV light and oxidation. This versatile tape is designed for permanent bonding in a variety of industrial and electronic applications.
Catalog No. 50-276-6678