Learn More
Grainger Double Sided Foam Tape 4 1/2 L PK40
Supplier:  Grainger 16U617
Encompass_Preferred
The 3M Double Sided Foam Tape is a tan colored double sided adhesive tape made by 3M. It has an acrylic adhesive and comes in multiple widths and lengths. The tape can bond many materials including metal, glass, plastic, and wood. It provides high strength bonding while allowing for movement due to its conformable foam core. The tape has excellent aging resistance while maintaining adhesion in temperatures from -20°F to 200°F. It can be used for mounting signs, attaching trim, and bonding components. The tape comes in multiple sizes ranging from 1/2 inch wide to 2 inches wide and lengths from 18 yards to 54 yards.
Catalog No. 50-276-6815