Learn More
Grainger Double Sided Foam Tape PK100
Supplier:  Grainger 16U623
Encompass_Preferred
The 3M Double Sided Foam Tape White is a high-performance acrylic adhesive tape with a polyethylene foam carrier. It has a thickness of 1/16 inch and is available in various widths and lengths. The tape features high initial tack and adhesion to most surfaces including plastics, glass, wood, and painted metal. It bonds on contact and offers stress relaxation to minimize lifting or curling. This versatile tape is ideal for mounting signs, nameplates, plastic parts, and trim. It can withstand temperatures up to 200°F for short periods. The tape has a shelf life of 18 months when stored at 72°F.
Catalog No. 50-276-6728