Learn More
Grainger Double Sided Foam Tape PK25
Supplier:  Grainger 16U778
Encompass_Preferred
The TAPECASE Double Sided Foam Tape is a white, double-sided adhesive tape made of polyethylene foam. It has an acrylic adhesive and is 0.045 inches thick with a peel adhesion of 40 oz/in. The tape can bond to most clean, dry surfaces including wood, glass, metal, plastic, and painted surfaces. It is useful for mounting signs, nameplates, moldings, and decorative trim. The tape resists UV rays and extreme temperatures. It comes in rolls that are 1 inch wide and 36 yards long.
Catalog No. 50-276-7033