Learn More
Grainger Double Sided Foam Tape PK25
Supplier:  Grainger 16U775
Encompass_Preferred
The TAPECASE Double Sided Foam Tape is a white, double-sided adhesive tape made of polyethylene foam. It has an acrylic adhesive and is 0.045 inches thick with a width of 1 inch. The tape can bond a variety of surfaces including wood, plastic, glass, and metal. It has high tack and is designed for permanent bonding for both indoor and outdoor applications. The tape resists UV rays, moisture, and extreme temperatures. It can be used for mounting signs, attaching molding or trim, and other bonding projects. The tape comes in a 36 yard roll and is made in the USA.
Catalog No. 50-276-6073