Learn More
Grainger Double Sided Foam Tape PK25
Supplier:  Grainger 16U774
Encompass_Preferred
The Grainger double-sided foam tape is available in various widths from 1/4" to 4" and lengths from 18 yards to 54 yards. It features an acrylic adhesive and has a maximum temperature resistance of 200°F. The tape has high shear and peel adhesion strength on most surfaces including wood, plastic, glass, and metal. It conforms to uneven surfaces and absorbs shock and vibration. The tape is designed for permanent bonding applications like trim molding, nameplates, plastic parts, and foam insulation. It has good resistance to UV rays, moisture, and chemicals. The tape can be die-cut into custom shapes.
Catalog No. 50-276-7239