Learn More
Grainger Double Sided Foam Tape PK25
Supplier:  Grainger 16U784
Encompass_Preferred
The TAPECASE Double Sided Foam Tape is a white, double-sided adhesive tape made of polyethylene foam. It has an acrylic adhesive and is 0.045 inches thick. The tape can be used for mounting, bonding, and splicing applications. It adheres well to wood, metal, glass, plastic, and painted surfaces. The tape is available in various widths and lengths up to 60 yards. It can withstand temperatures from -20°F to 200°F. The tape has high shear and peel adhesion strength.
Catalog No. 50-276-4998