Learn More
Grainger Double Sided Foam Tape PK25
Supplier:  Grainger 16U781
Encompass_Preferred
The Grainger website features 3M VHB double-sided foam mounting tape. This high-performance acrylic adhesive tape provides a very high bond strength and is offered in various thicknesses, widths and lengths. It bonds well to high and medium surface energy substrates like metal, glass and plastic. The tape is designed for permanent bonding applications and can replace rivets, spot welds, liquid adhesives and other permanent fasteners. It resists solvents, moisture and plasticizer migration. The foam core provides stress relaxation and conforms to uneven surfaces. Overall, this is a versatile, heavy-duty double-sided foam tape ideal for permanent bonding and mounting applications.
Catalog No. 50-276-4888