Learn More
Grainger Double Sided Foam Tape PK25
Supplier:  Grainger 16U782
Encompass_Preferred
The TAPECASE Double Sided Foam Tape is a white, double-sided adhesive tape made of polyethylene foam. It has an acrylic adhesive and is 0.045 inches thick with a width of 1 inch. The tape can bond a variety of materials including paper, plastic, glass, and metal. It has high tack and is designed for permanent bonding for crafts, scrapbooking, mounting, and general repairs. The tape resists UV rays, moisture, and extreme temperatures. It can be easily cut to desired lengths.
Catalog No. 50-276-4889