Learn More
Grainger Double Sided Foam Tape PK25
Supplier:  Grainger 16U779
Encompass_Preferred
The TAPECASE Double Sided Foam Tape is a white, double-sided adhesive tape made of polyethylene foam. It has an acrylic adhesive and is 0.045 inches thick with a width of 1 inch. The tape can bond a variety of surfaces including wood, plastic, glass, and metal. It has high tack and is designed for permanent bonding for both indoor and outdoor applications. The operating temperature range is -4°F to 176°F. It comes in a 36 yard roll and can be easily cut to desired lengths. The tape provides good conformability and shock absorption.
Catalog No. 50-276-7263