Learn More
Grainger Double Sided Foam Tape PK25
Supplier:  Grainger 16U776
Encompass_Preferred
The TapeCase Double Sided Foam Tape is a white, double-sided adhesive tape made of polyethylene foam. It has an acrylic adhesive and is 0.045 inches thick. The tape can be used for mounting, bonding, and attaching applications. It adheres well to most clean, dry surfaces including wood, glass, metal, plastic, and painted surfaces. The tape is sold in rolls that are 1 inch wide and 36 yards long. It can withstand temperatures up to 200°F. The tape provides a strong, permanent bond while also absorbing vibration and impact. It conforms well to uneven surfaces.
Catalog No. 50-276-5375