Learn More
Grainger Double Sided Foam Tape PK27
Supplier:  Grainger 16U618
Encompass_Preferred
The 3M Scotch Double Sided Tape is a high performance adhesive tape that has a natural rubber adhesive on both sides. It comes in a pack of 27 rolls that are 1 inch wide x 60 yards long. The tape is designed for permanent bonding and mounting applications. It bonds well to paper, cardboard, wood, glass, and metal. The tape has good temperature resistance, aging stability, and solvent resistance. It can be used for packaging, splicing, attaching signs or nameplates, and general repair.
Catalog No. 50-276-6816