Learn More
ARCTICZYMES Double-strand specific Dnases
Supplier:  ARCTICZYMES 70600201
Encompass
Double-strand specific DNases (dsDNases) are unique double-strand specific endonucleases. As they do not digest ssDNA or RNA, they can be used to specifically remove dsDNA in the presence of other nucleic acids. The enzymes are heat-labile, which makes them ideal for applications where the DNase have to be inactivated. There are two versions of this enzyme available, dsDNase and HL-dsDNase.
Catalog No. NC1772349