Learn More
Analytical Sales & Services 60cm Double UHPLC Assembly - 1/16” OD x 0.005” ID
Supplier:  Analytical Sales & Services 62560
Encompass
60cm x 0.005¿ID Ultra High Pressure HPLC Fitting Assemblies are the perfect column connectorsfor HPLC because they totally eliminate any possibility of Dead Volume in thecritical inlet and outlet areas, while holding pressure to 27,000psi.
Catalog No. NC0006498