Learn More
Abcam DoubleStain IHC Kit: G&M on human tissue (Green/HRP & AP/Red), 18ML
Supplier:  Abcam AB18327218ML
Encompass_Preferred
Abcam's DoubleStain IHC kit (ab183272) is designed to use with user supplied goat and mouse antibodies to detect two distinct antigens on human tissue or cell samples.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-267-3167