Learn More
Sigma Aldrich Fine Chemicals Biosciences Dounce tissue grinder tube1EA
Supplier:  Sigma Aldrich Fine Chemicals Biosciences T25671EA
Encompass
Designed primarily for cellular work where the nucleus remains intact after homogenization.
Catalog No. 50-290-1888